Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Women's Classic Heavyweight Excel FR™ Flame Resistant Jean, Blue Denim

Catalog No. 17200732 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
4
6
8
10
12
14
16
18
20
22
24
Size (English) Inseam:
28 in.
30 in.
32 in.
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Inseam
17-200-732 12 28 in.
17-200-720 4 28 in.
17-200-721 4 30 in.
17-200-722 4 32 in.
17-200-723 6 28 in.
17-200-726 8 28 in.
17-200-729 10 28 in.
17-200-724 6 30 in.
17-200-727 8 30 in.
17-200-725 6 32 in.
17-200-728 8 32 in.
17-200-731 10 32 in.
17-200-730 10 30 in.
17-200-736 14 30 in.
17-200-733 12 30 in.
17-200-734 12 32 in.
17-200-735 14 28 in.
17-200-738 16 28 in.
17-200-737 14 32 in.
17-200-739 16 30 in.
17-200-740 16 32 in.
17-200-741 18 28 in.
17-200-742 18 30 in.
17-200-743 18 32 in.
17-200-744 20 28 in.
17-200-746 20 32 in.
17-200-747 22 28 in.
17-200-745 20 30 in.
17-200-748 22 30 in.
17-200-750 24 28 in.
17-200-749 22 32 in.
17-200-751 24 30 in.
17-200-752 24 32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
This item is not returnable. View return policy
Catalog No. 17-200-732 Supplier Bulwark™ FR Supplier No. PEJ3DW 12 28
This item is not returnable. View return policy

This heavyweight CAT 2 AR/FR Denim Jean is NFPA™ 2112 compliant. It offers a classic workwear look and fit with FR protection, designed specifically with women in mind. Classic fit sits at the natural waist and has a slimmer leg opening that will fit over boots.

  • Primary Closure: Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
  • Blend: 100% Pre-Washed Cotton Denim
  • Closure: One-piece waistband with button closure
  • Pocket: Five jean-style pockets
  • Features:
    • One-piece waistband with button closure
    • Five jean-style pockets
    • Two-needle felled seam construction and soft pre-wash for comfort and look
    • Classic fit is traditional, sits at the natural waist and has a slimmer leg opening that will fit over boots
  • Product Collection: Excel FR
  • Protection: ATPV Arc Rating: 21
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
  • Gender: Female
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 12
Gender Women's
Product Type Flame-resistant Jeans
Material Cotton
Color Blue
No. of Pockets 5
Closure Type Button
Hazard Risk Category HRC2
Arc Flash Rating ATPV 20.7 Calories/cm2
Certifications/Compliance Meets NFPA 2112, UL
Size (English) Inseam 28 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.