Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Lightweight Nomex™ Flame Resistant Uniform Shirt

Catalog No. 191661035 Shop All Red Kap Industries Products
Change view
Click to view available options
Color:
Navy
Royal Blue
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Long)
Medium (Regular)
Small (Regular)
X-Large (Long)
X-Large (Regular)
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-166-1035 Royal Blue Large (Long)
19-166-1018 Navy Small (Regular)
19-166-1019 Navy Medium (Regular)
19-166-1020 Navy Large (Regular)
19-166-1021 Navy X-Large (Regular)
19-166-1024 Navy Medium (Long)
19-166-1023 Navy 3X-Large (Regular)
19-166-1026 Navy X-Large (Long)
19-166-1027 Navy 2X-Large (Long)
19-166-1025 Navy Large (Long)
19-166-1022 Navy 2X-Large (Regular)
19-166-1028 Royal Blue Small (Regular)
19-166-1029 Royal Blue Medium (Regular)
19-166-1031 Royal Blue X-Large (Regular)
19-166-1033 Royal Blue 3X-Large (Regular)
19-166-1030 Royal Blue Large (Regular)
19-166-1032 Royal Blue 2X-Large (Regular)
19-166-1034 Royal Blue Medium (Long)
19-166-1036 Royal Blue X-Large (Long)
19-166-1037 Royal Blue 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Catalog No. 19-166-1035 Supplier Bulwark™ FR Supplier No. SND2RB LN L
Add to Cart
Add to Cart
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.

This Lightweight FR Uniform Shirt offers a tailored, professional look with CAT 1 AR/FR protection and NFPA™ 2112 compliance. Tailored sleeve placket, topstitched cuffs and straight back yoke look sharp and professional while being protective.

  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 1 Protection
  • Protection: Arc Rating ATPV 4.8 calories/cm2
  • Hazard: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 4.5 oz. (150 g/m2) Nomex™ IIIA
  • Blend: 93% Nomex™ Aramid/5% Kevlar™ Aramid/2% Other Fiber
  • Closure: Button-down front and topstitched cuff with button closure
  • Collar: Banded topstitched collar
  • Pocket: Two chest pockets with button flap closures and sewn in pocket stall
  • Placket front with button closure
  • Tailored sleeve placket
  • Product Collection: Nomex™
  • Protection: ATPV Arc Rating: 4.4
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Nomex™
  • Brand: Bulwark™

Specifications

Size Large (Long)
Gender Men's
Material Nomex IIIA
Color Royal Blue
Closure Type Button
Hazard Risk Category 1
Arc Flash Rating ATPV 4.8 Calories/cm2
Certifications/Compliance NFPA 2112, NFPA 70E, Category 1 Protection
Weight (English) 4.5 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.