Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Lightweight Nomex™ Flame Resistant Uniform Shirt

Catalog No. 191661020
Click to view available options
Color:
Navy
Royal Blue
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Long)
Medium (Regular)
Small (Regular)
X-Large (Long)
X-Large (Regular)
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.

This Lightweight FR Uniform Shirt offers a tailored, professional look with CAT 1 AR/FR protection and NFPA™ 2112 compliance. Tailored sleeve placket, topstitched cuffs and straight back yoke look sharp and professional while being protective.

  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 1 Protection
  • Protection: Arc Rating ATPV 4.8 calories/cm2
  • Hazard: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 4.5 oz. (150 g/m2) Nomex™ IIIA
  • Blend: 93% Nomex™ Aramid/5% Kevlar™ Aramid/2% Other Fiber
  • Closure: Button-down front and topstitched cuff with button closure
  • Collar: Banded topstitched collar
  • Pocket: Two chest pockets with button flap closures and sewn in pocket stall
  • Placket front with button closure
  • Tailored sleeve placket
  • Product Collection: Nomex™
  • Protection: ATPV Arc Rating: 4.4
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Nomex™
  • Brand: Bulwark™

Specifications

Size Large (Regular)
Gender Men's
Material Nomex IIIA
Color Navy
Closure Type Button
Hazard Risk Category 1
Arc Flash Rating ATPV 4.8 Calories/cm2
Certifications/Compliance NFPA 2112, NFPA 70E, Category 1 Protection
Weight (English) 4.5 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.