Learn More
Bulwark™ FR Nomex™ IIIA HRC1 6 oz. Uniform Shirts
Click to view available options
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Long)
Medium (Regular)
X-Large (Long)
X-Large (Regular)
Description
- Flame-resistant, 6 oz. Nomex IIIA
- Banded, topstitched collar
- Topstitched cuffs with adjustable button closures
- Placket front with button closure
- Tailored sleeve placket
Specifications
Specifications
Size | 3X-Large (Regular) |
Material | Nomex IIIA |
Color | Royal Blue |
Hazard Risk Category | 1 |
Weight (English) | 6 oz. |