Learn More
Bulwark™ FR Men's Heavyweight Excel FR™ Flame Resistant Denim Dungaree, Stonewash
Shop All Red Kap Industries ProductsDescription
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCELFR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Cotton Denim
- Closure: Zipper fly with single metal button on waistband
- Pocket: Two deep scoop pockets, Two oversized reinforced hip pockets, One rule pocket on right leg with wrap around utility pocket
- Features:
- Sits at natural waist
- Loose seat and thigh with wider leg opening
- Two deep scoop pockets and two oversized reinforced hip pockets with rule pocket on right leg and wrap around utility pocket
- Soft pre-wash for comfort and look
- Product Collection: Excel FR, Denim
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at the waist. Extra room in the seat and thigh. Relaxed in the leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
| Size | 34 x 32 in. |
| Gender | Men's |
| Product Type | Flame-resistant Dungarees |
| Material | Cotton |
| Color | Blue |
| No. of Pockets | 5 |
| Closure Type | Concealed button |
| Resistant To | Flame |
| Hazard Risk Category | HRC2 |
| Arc Flash Rating | ATPV 20.7 Calories/cm2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.