Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Heavyweight Excel FR™ Flame Resistant Denim Dungaree, Stonewash

Catalog No. 19296514 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-296-514 38 x 34 in.
19-296-500 30 x 30 in.
19-296-502 30 x 34 in.
19-296-501 30 x 32 in.
19-296-503 32 x 30 in.
19-296-504 32 x 32 in.
19-296-507 34 x 32 in.
19-296-506 34 x 30 in.
19-296-505 32 x 34 in.
19-296-508 34 x 34 in.
19-296-510 36 x 32 in.
19-296-509 36 x 30 in.
19-296-511 36 x 34 in.
19-296-512 38 x 30 in.
19-296-513 38 x 32 in.
19-296-515 40 x 30 in.
19-296-517 40 x 34 in.
19-296-516 40 x 32 in.
19-296-518 42 x 30 in.
19-296-519 42 x 32 in.
19-296-520 42 x 34 in.
19-296-522 44 x 32 in.
19-296-521 44 x 30 in.
19-296-523 44 x 34 in.
19-296-525 46 x 32 in.
19-296-527 48 x 30 in.
19-296-524 46 x 30 in.
19-296-526 46 x 34 in.
19-296-528 48 x 32 in.
19-296-530 50 x 30 in.
19-296-529 48 x 34 in.
19-296-531 50 x 32 in.
19-296-532 50 x 34 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
This item is not returnable. View return policy
Catalog No. 19-296-514 Supplier Bulwark™ FR Supplier No. PEJ8SW 38 34
Add to Cart
Add to Cart
This item is not returnable. View return policy

This heavyweight CAT 2 AR/FR dungaree style denim jean is NFPA™ 2112 compliant. Convenient features like a hammer loop on left leg and utility loop on waistband ensure you've got a dedicated place for all your tools. Loose fit improves comfort and a wider leg opening fits easily over boots.

  • Primary Closure: Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: EXCELFR™ Flame resistant, 14.75 oz. (500 g/m2)
  • Blend: 100% Cotton Denim
  • Closure: Zipper fly with single metal button on waistband
  • Pocket: Two deep scoop pockets, Two oversized reinforced hip pockets, One rule pocket on right leg with wrap around utility pocket
  • Features:
    • Sits at natural waist
    • Loose seat and thigh with wider leg opening
    • Two deep scoop pockets and two oversized reinforced hip pockets with rule pocket on right leg and wrap around utility pocket
    • Soft pre-wash for comfort and look
  • Product Collection: Excel FR, Denim
  • Protection: ATPV Arc Rating: 21
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Extra room in the seat and thigh. Relaxed in the leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 38 x 34 in.
Product Type Flame-resistant Dungarees
Gender Men's
Material Cotton
Color Blue
No. of Pockets 5
Closure Type Concealed button
Resistant To Flame
Hazard Risk Category HRC2
Arc Flash Rating ATPV 20.7 Calories/cm2
Size (English) Inseam 34 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.