Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Loose Flame Resistant Jean, Stonewash

Catalog No. 19296655 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
28 x 30 in.
28 x 32 in.
28 x 34 in.
30 x 30 in.
30 x 32 in.
30 x 34 in.
31 x 30 in.
31 x 32 in.
31 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
45 product options available for selection
Product selection table with 45 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-296-655 40 x 30 in.
19-296-628 28 x 30 in.
19-296-630 28 x 34 in.
19-296-629 28 x 32 in.
19-296-631 30 x 30 in.
19-296-635 31 x 32 in.
19-296-633 30 x 34 in.
19-296-634 31 x 30 in.
19-296-632 30 x 32 in.
19-296-636 31 x 34 in.
19-296-638 32 x 32 in.
19-296-637 32 x 30 in.
19-296-639 32 x 34 in.
19-296-640 33 x 30 in.
19-296-642 33 x 34 in.
19-296-643 34 x 30 in.
19-296-644 34 x 32 in.
19-296-641 33 x 32 in.
19-296-646 35 x 30 in.
19-296-645 34 x 34 in.
19-296-647 35 x 32 in.
19-296-648 35 x 34 in.
19-296-649 36 x 30 in.
19-296-652 38 x 30 in.
19-296-650 36 x 32 in.
19-296-651 36 x 34 in.
19-296-653 38 x 32 in.
19-296-654 38 x 34 in.
19-296-656 40 x 32 in.
19-296-657 40 x 34 in.
19-296-658 42 x 30 in.
19-296-661 44 x 30 in.
19-296-663 44 x 34 in.
19-296-662 44 x 32 in.
19-296-659 42 x 32 in.
19-296-664 46 x 30 in.
19-296-660 42 x 34 in.
19-296-665 46 x 32 in.
19-296-666 46 x 34 in.
19-296-667 48 x 30 in.
19-296-669 48 x 34 in.
19-296-671 50 x 32 in.
19-296-668 48 x 32 in.
19-296-670 50 x 30 in.
19-296-672 50 x 34 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
45 options
This item is not returnable. View return policy
Catalog No. 19-296-655 Supplier Bulwark™ FR Supplier No. PEJ6SW 40 30
Add to Cart
Add to Cart
This item is not returnable. View return policy

This heavyweight CAT 2 AR/FR denim jean is NFPA™ 2112 compliant. With classic five-pocket styling, the jean offers a loose fit for easy comfort with FR protection. New, stone-washed fabric is pre-washed for greater comfort and look, while a wider leg opening fits easily over boots.

  • Primary Closure: Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
  • Blend: 100% Pre-Washed Cotton Denim
  • Closure: One-piece waistband with metal button closure (metal is not exposed to the skin)
  • Pocket: Five jean-style pockets
  • Features:
    • Sits below natural waist
    • Loose seat and thigh with wider leg opening than our classic fit jean
    • Soft pre-wash and stone-washed fabric for comfort and look
  • Product Collection: Excel FR, Denim
  • Packaging: Waistband ticket
  • Protection: ATPV Arc Rating: 21
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits just below the waist. Extra room in the seat and thigh. Fuller in the leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 40 x 30 in.
Gender Men's
Product Type Flame-resistant Jeans
Material Denim
Color Blue
No. of Pockets 5
Closure Type Zipper
Resistant To Flame
Hazard Risk Category HRC2
Size (English) Inseam 30 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.