Learn More
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Pre-Washed Cotton Denim
- Closure: One-piece waistband with metal button closure (metal is not exposed to the skin)
- Pocket: Five jean-style pockets
- Features:
- Sits below natural waist
- Loose seat and thigh with wider leg opening than our classic fit jean
- Soft pre-wash and stone-washed fabric for comfort and look
- Product Collection: Excel FR, Denim
- Packaging: Waistband ticket
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits just below the waist. Extra room in the seat and thigh. Fuller in the leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
| Size | 44 x 32 in. |
| Product Type | Flame-resistant Jeans |
| Gender | Men's |
| Material | Denim |
| Color | Blue |
| No. of Pockets | 5 |
| Closure Type | Zipper |
| Resistant To | Flame |
| Hazard Risk Category | HRC2 |
| Arc Flash Rating | ATPV 20.7 Calories/cm2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.