Learn More
VF Workwear Bulwark Women's Flame Resistant Curvy Fit Denim Jeans
Shop All Red Kap Industries ProductsSpecifications
Specifications
| Size | 8 |
| Gender | Women's |
| Material | Cotton, Modacrylic |
| Color | Blue |
| No. of Pockets | 5 |
| Closure Type | Button |
| Hazard Risk Category | HRC2 |
| Arc Flash Rating | EBT 13.0 Calories/cm2 |
| Size (English) Inseam | 34 in. |
| Style | Unhemmed |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.