Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bumetanide, 98+%, PROMO, Thermo Scientific Chemicals

Catalog No. J6230206PM
Click to view available options
Quantity:
5 g

Inhibitor of the sodium-potassium-chloride cotransporter

Applications

Bumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1

Solubility

Soluble in ethanol (10 mg/mL), DMSO (25 mg/mL), acetone, benzene, methanol, propylene glycol, and water (<1mg/mL).

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28395-03-1
Molecular Formula C17H20N2O5S
Molecular Weight (g/mol) 364.42 g/mol
MDL Number MFCD00078949
InChI Key MAEIEVLCKWDQJH-UHFFFAOYSA-N
Synonym bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum
PubChem CID 2471
ChEBI CHEBI:3213
IUPAC Name 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid
SMILES CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2

Specifications

Melting Point 230°C to 231°C
Color White
Quantity 5 g
Merck Index 14,1484
Solubility Information Soluble in ethanol, DMSO, acetone, benzene, methanol, propylene glycol, and water.
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Bumetanide
EINECSNumber 249-004-6
RTECSNumber DG4910000
TSCA No

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.