Learn More
United Scientific™ Bumper Guard For Volumetric Flask

Shop All United Scientific Supplies, Inc Products


Description
- Silicone bumpers slip easily onto any volumetric flask. In the event of a flask being accidentally tipped over, the wide bumpers cushion the neck from hitting the table surface.
- Color coded and available in five different sizes.
- Wide design to cushion flasks if tipped over
- Made of high-quality silicone
Specifications
Specifications
| Capacity (Metric) Media | 500 mL |
| For Use With (Equipment) | Volumetric Flask |
| Product Type | Bumper Guard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.