Learn More
Bupivacaine
Description
Bupivacaine acts as a potent and long-acting local anesthetic agent in vivo. At 100 μM, it has been reported to inhibit the uptake of dopamine in striatal synaptosomes by 47%.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Application
- Bupivacaine has been shown to activate the AMPKα/BDNF signaling pathway in spinal astrocytes

Specifications
Specifications
Melting Point | 106°C to 110°C |
Color | White |
Quantity | 1 g |
UN Number | UN2811 |
Solubility Information | Soluble in water,ethanol,chloroform (slightly ),and acetone (slightly ). |
Formula Weight | 288.43 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Bupivacaine |
Safety and Handling
Hazard Category | H300+H310+H330 |
Hazard Statement | GHS H Statement H300-H310-H330 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. |
Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 253-911-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.