Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupivacaine hydrochloride monohydrate, 98+%

Catalog No. AAJ6283506
Click to view available options
Quantity:
1 g
5 g
25 g

Inhibits TREK-1 channels and depolarizes the cell membrane | CAS: 73360-54-0 | C18H31ClN2O2 | 342.908 g/mol

It is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.

Solubility
Soluble in water to 50 mM, in DMSO to 100 mM and in ethanol to 100 mM.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73360-54-0
Molecular Formula C18H31ClN2O2
Molecular Weight (g/mol) 342.908
MDL Number MFCD00078956
InChI Key HUCIWBPMHXGLFM-UHFFFAOYSA-N
Synonym bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate
PubChem CID 5282419
IUPAC Name 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride
SMILES CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl

Specifications

Melting Point 255°C to 259°C
Color White
Quantity 5 g
UN Number UN2811
Merck Index 14,1495
Solubility Information Soluble in water to 50 mM,in DMSO to 100 mM and in ethanol to 100 mM.
Formula Weight 342.9
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Bupivacaine hydrochloride monohydrate
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 241-917-8
RTECSNumber TK6125000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.