Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupivacaine hydrochloride monohydrate, ≥99%, MilliporeSigma™ Supelco™

$88.80 - $356.50

Specifications

Percent Purity ≥99%
Grade Analytical Standard (for drug analysis)
Linear Formula C18H28N2 O · HCl · H2O
Physical Form Neat
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9313
View Documents
MilliporeSigma Supelco
B52741G
1 g
Each for $88.80
Add to cart
 
11-101-8747
View Documents
MilliporeSigma Supelco
B52745G
5 g
Each for $150.30
Add to cart
 
11-101-8210
View Documents
MilliporeSigma Supelco
B527415G
15 g
Each for $356.50
Add to cart
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

≥99%
Analytical Standard (for drug analysis)
C18H28N2 O · HCl · H2O
Neat
Safety and Handling

Safety and Handling

RTECSNumber : TK6125000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit