Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buprenorphine Mouse, Unconjugated, Clone: HM376, Abnova™

Catalog No. 89339266 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-266 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-266 Supplier Abnova Corporation Supplier No. MAB21880
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Buprenorphine Monoclonal antibody specifically detects Buprenorphine in samples. It is validated for ELISA, LFIA.

Specifications

Antigen Buprenorphine
Applications ELISA, Lateral Flow
Classification Monoclonal
Clone HM376
Conjugate Unconjugated
Host Species Mouse
Immunogen Buprenorphine conjugated with BSA
Quantity 1 mg
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.