Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupropion Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601135
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

B1237, 31677-93-7, C13H18ClNO·HCl

Spectrum™ Chemical Bupropion Hydrochloride, USP is an antidepressant medication. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 31677-93-7
Molecular Formula C13H19Cl2NO
Molecular Weight (g/mol) 276.20
InChI Key HEYVINCGKDONRU-UHFFFAOYNA-N
IUPAC Name hydrogen 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one chloride
SMILES [H+].[Cl-].CC(NC(C)(C)C)C(=O)C1=CC=CC(Cl)=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98-102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.