Learn More
United Scientific™ Burette Clamp
SDPSCI_ED
Catalog No. S15900 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Type:
Burette Clamp (Double)
Catalog No. Type
S15900 Burette Clamp (Double)
1 options
Catalog No. S15900 Supplier United Scientific™ Supplier No. 45102
Only null left
Add to Cart
Add to Cart
Sturdy Clamp that can be used with virtually all support stands and lattice setups.
  • These polypropylene burette clamps are completely corrosion-proof.
  • The spring-loaded jack in the middle provides a vice like grip on the burette and prevents it from slipping.
  • Two tapered wedges are included to ensure firm grip on 1/2 or 3/8 in. diameter rods.
Specifications
Type Burette Clamp (Double)
Material Polypropylene
Color White
Jaw Type Spring Loaded