Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Burkholderia multivorans derived from ATCC™ BAA-247™
SDP

Catalog No. p-8707011 Shop All Microbiologics Products
Click to view available options
Quantity:
2 Pack
6 Pack
This item is not returnable. View return policy
This item is not returnable. View return policy

Simple, all-inclusive design making life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab.

  • Self-contained units of a single organism

Specifications

ATCC® ATCC™ BAA-247
Biosafety Level 2
Content And Storage 2°C to 8°C
Format KWIK-STIK™
Manufacturing Origin United States
For Use With (Application) Microbial Identification
Packaging Type Plastic Canister
Quantity 2 Pack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.