Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Burn-up Bin™ Biohazard Waste Boxes
SCI_ED

Catalog No. s37098 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W x H):
8 x 8 x 10 in.
This item is not returnable. View return policy

Catalog No. S37098

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Store flat, assemble in seconds.

  • Corrugated cardboard—single use
  • 2mil polyliner safely holds biohazard waste
  • Tight-fitting lid has flap opening to accept material; to dispose, simply close lid and transfer filled box to incinerator
  • Graphics and biohazard symbols readily identify contents, encourage proper use

Specifications

Design Bench model
Diameter (Metric) Outer 17.8 cm
Diameter (English) Outer 7 in.
Dimensions (L x W x H) 8 x 8 x 10 in.
Height (Metric) 25 cm
Length (Metric) 20 cm
Width (Metric) 20 cm
Liner Material Polyethylene
For Use With (Application) Quick identification for non-hazardous waste paper products
Width (English) 8 in.
Length (English) 8 in.
Thickness 2 mm
Height (English) 10 in.
Product Type Benchtop Biohazard Waste Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.