Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buspirone hydrochloride

Catalog No. aaj6047603
Click to view available options
Quantity:
1 g
5 g

CAS: 33386-08-2 | C21H32ClN5O2 | 421.97 g/mol

It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

Solubility
Soluble in methanol (50 mg/ml), water (partly), DMSO (100 mM).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33386-08-2
Molecular Formula C21H32ClN5O2
Molecular Weight (g/mol) 421.97
MDL Number MFCD00078569
InChI Key RICLFGYGYQXUFH-UHFFFAOYSA-N
Synonym buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
PubChem CID 36431
ChEBI CHEBI:3224
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
SMILES C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl

Specifications

Color White
Quantity 1 g
UN Number UN2811
Merck Index 14,1504
Solubility Information Soluble in methanol (50mg/ml),water (partly),DMSO (100 mM).
Formula Weight 421.96
Physical Form Powder
Chemical Name or Material Buspirone hydrochloride
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 251-489-4
RTECSNumber CL9915000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.