Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butenafine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B32931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32931G 1 g
B32935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32931G Supplier TCI America Supplier No. B32931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 101827-46-7
Molecular Formula C23H28ClN
Molecular Weight (g/mol) 353.934
MDL Number MFCD00917064
InChI Key LJBSAUIFGPSHCN-UHFFFAOYSA-N
PubChem CID 443867
ChEBI CHEBI:31325
IUPAC Name 1-(4-tert-butylphenyl)-N-methyl-N-(naphthalen-1-ylmethyl)methanamine;hydrochloride
SMILES CC(C)(C)C1=CC=C(C=C1)CN(C)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

Melting Point 210°C
Color White
Quantity 1 g
Formula Weight 353.93
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butenafine Hydrochloride
RTECSNumber QJ8585000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.