Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl acrylate, 99+%, stabilized, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
141-32-2,150-76-5
C7H12O2
128.17
MFCD00009446
CQEYYJKEWSMYFG-UHFFFAOYSA-N
butyl acrylate,n-butyl acrylate,acrylic acid butyl ester,n-butyl propenoate,2-propenoic acid, butyl ester,butyl 2-propenoate,butylacrylate,acrylic acid, butyl ester,acrylic acid n-butyl ester,butylester kyseliny akrylove
8846
CHEBI:3245
butyl prop-2-enoate
CCCCOC(=O)C=C
141-32-2,150-76-5
C7H12O2
128.17
MFCD00009446
CQEYYJKEWSMYFG-UHFFFAOYSA-N
butyl acrylate,n-butyl acrylate,acrylic acid butyl ester,n-butyl propenoate,2-propenoic acid, butyl ester,butyl 2-propenoate,butylacrylate,acrylic acid, butyl ester,acrylic acid n-butyl ester,butylester kyseliny akrylove
8846
CHEBI:3245
butyl prop-2-enoate
CCCCOC(=O)C=C
Color Undesignated
Viscosity 0.869 mPa.s (20°C)
Refractive Index 1.4170 to 1.4190
Flash Point 39°C
Infrared Spectrum Authentic
Specific Gravity 0.89
Packaging Glass Bottle
Stabilizer 10 to 20ppm MEHQ
Melting Point -64.0°C
Boiling Point 145.0°C
Quantity 2.5L
Density 0.8900g/mL
Acidity 0.01% max. (as acrylic acid)
Assay Percent Range 99% min. (GC)
Linear Formula H2C=CHCO2(CH2)3CH3
Beilstein 02, II, 388
Merck Index 15, 1541
Solubility Information Solubility in water: 1.4g/L (20°C)
Formula Weight 128.17
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Butyl acrylate
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful to aquatic li

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f

GHS Signal Word: Warning

EINECSNumber : 205-480-7