Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl benzoate, 98+%

Catalog No. AC296400010
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296400010 1 kg Glass bottle
AC296400050 5 g Glass bottle
AC296402500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC296400010 Supplier Thermo Scientific Chemicals Supplier No. 296400010
Add to Cart

CAS: 136-60-7 | C11H14O2 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-60-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00009439
InChI Key XSIFPSYPOVKYCO-UHFFFAOYSA-N
Synonym n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
PubChem CID 8698
IUPAC Name butyl benzoate
SMILES CCCCOC(=O)C1=CC=CC=C1

Specifications

Melting Point -22°C
Color Yellow
Density 1.0000g/mL
Boiling Point 250°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4960 to 1.498
Linear Formula C6H5COO(CH2)3CH3
Quantity 1 kg
Specific Gravity 1
Merck Index 15, 1554
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 178.23
Percent Purity 98+%
Physical Form Oily Liquid
Chemical Name or Material Butyl benzoate, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 205-252-7
RTECSNumber DG4925000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.