Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl glycidyl ether, 95%, Thermo Scientific™

Catalog No. AC438072500
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-11-3
Molecular Formula C18H14O
Molecular Weight (g/mol) 246.31
InChI Key ATGFTMUSEPZNJD-UHFFFAOYSA-N
Synonym 1,1':3',1-terphenyl-2'-ol, m-terphenyl-2'-ol, 2,6-diphenyl phenol, 2'-hydroxy-m-terphenyl, unii-la93w35m96, 3-phenyl-1,1'-biphenyl-2-ol, 2,6-di-phenylphenol, 2,6,-diphenylphenol, 2,6-di phenyl phenol
PubChem CID 75512
IUPAC Name 2,6-diphenylphenol
SMILES C1=CC=C(C=C1)C2=C(C(=CC=C2)C3=CC=CC=C3)O

Specifications

Color Colorless
Density 0.9200g/mL
Boiling Point 167.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4170 to 1.4190 (20°C, 589nm)
Quantity 250 mL
Formula Weight 130.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Butyl glycidyl ether
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects. Harmful if inhaled. Harmful if swallowed. May cause respiratory irritation. May cause an allergic skin reaction. Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.