Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Trifluoromethanesulfonate, TCI America™
SDP

Catalog No. T203425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T203425G 25 g
T20345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T203425G Supplier TCI America Supplier No. T203425G

Chemical Identifiers

CAS 75618-25-6
Molecular Formula C5H9F3O3S
Molecular Weight (g/mol) 206.179
MDL Number MFCD00799500
InChI Key SEXLAQXMAFCJCO-UHFFFAOYSA-N
Synonym Butyl Triflate, Trifluoromethanesulfonic Acid Butyl Ester
PubChem CID 10921660
IUPAC Name butyl trifluoromethanesulfonate
SMILES CCCCOS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 50°C
UN Number 2924
Quantity 25 g
Formula Weight 206.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.