Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylated hydroxyanisole, 96%

Catalog No. p-79063
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 25013-16-5 | C11H16O2 | 180.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.24
MDL Number MFCD01779059
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
Synonym 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
PubChem CID 8456
ChEBI CHEBI:76358
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=C(C=CC(=C1)OC)O

Specifications

Melting Point 48°C to 55°C
Color White to Yellow
Boiling Point 268°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Merck Index 15,1549
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,propylene glycol,,chlorogorm,ether and acetone,slightly soluble in benzene
Formula Weight 180.24
Percent Purity 96%
Physical Form Crystalline Flakes or Crystals
Chemical Name or Material Butylated hydroxyanisole, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED:
EINECSNumber 246-563-8
RTECSNumber SL1945000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.