Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylbenzene, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019075 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mL

Butylbenzene is a colourless organic solvent. It is used in pesticide manufacturing, plasticizer and also as solvent for coating. It is sometimes used as surface active agent, polymer linking agent and in organic synthesis.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 104-51-8
Ignition Point 774°F
Vapor Pressure 1.03 mmHg (23°C)
Vapor Density >1 (vs. air)
Density 0.86 g/mL (at 25°C)
Flash Point 59°C
Physical Form Neat
Linear Formula C6H5(CH2)3 CH3
MDL Number MFCD00009463
Melting Point -88°C
Boiling Point 183°C
Synonym 1-Phenylbutane
Type Analytical Standard
Quantity 10 mL
Formula Weight 134.22
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.