Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylboronic Acid (contains varying amounts of Anhydride) [for Esterification], TCI America™
SDP

Catalog No. B052925G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4426-47-5
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00002106
InChI Key QPKFVRWIISEVCW-UHFFFAOYSA-N
Synonym n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl
PubChem CID 20479
IUPAC Name butylboronic acid
SMILES B(CCCC)(O)O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 25 g
Formula Weight 101.94
Physical Form Crystalline Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.