Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylmalonic Acid 98.0+%, TCI America™
SDP

Catalog No. B441425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B441425G 25 g
B44145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B441425G Supplier TCI America Supplier No. B441425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 534-59-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00002673
InChI Key MCRZWYDXIGCFKO-UHFFFAOYSA-N
PubChem CID 10801
IUPAC Name 2-butylpropanedioic acid
SMILES CCCCC(C(=O)O)C(=O)O

Specifications

Melting Point 103°C
Color White
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butylmalonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.