Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Butyric Acid
SCI_ED

Catalog No. p-4416674 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Lab Grade
Ester synthesis

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
Synonym butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
PubChem CID 264
ChEBI CHEBI:30772
IUPAC Name butanoic acid
SMILES CCCC(O)=O

Specifications

Color Colorless
Assay Percent Range >99%
Linear Formula CH3CH2CH2COOH
Formula Weight 88.11
Grade Laboratory
Density 0.9640g/cmsuperscript_3;
Quantity 100 mL
Packaging Lab Grade
Chemical Name or Material Butyric Acid
ShelfLife 5 years
DOTInformation DOT Class 8, III, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.