Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyric acid hydrazide, 95%, Thermo Scientific™

Catalog No. AC147460250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC147460250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC147460250 Supplier Thermo Scientific Chemicals Supplier No. 147460250

Chemical Identifiers

CAS 3538-65-6
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
InChI Key FCCCRBDJBTVFSJ-UHFFFAOYSA-N
Synonym butyrohydrazide, butyric acid hydrazide, butanohydrazide, butyrylhydrazine, butanoic acid, hydrazide, n-butyric acid hydrazide, butyric hydrazide, butryic acid hydrazide, butanoic acid hydrazide, butyryl hydrazine
PubChem CID 77079
IUPAC Name butanehydrazide
SMILES CCCC(=O)NN

Specifications

Melting Point 43.0°C to 44.0°C
Boiling Point 120.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min
Linear Formula CH3CH2CH2C(O)NHNH2
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 102.14
Percent Purity 95%
Chemical Name or Material Butyric acid hydrazide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 222-579-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.