Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyrophenone, 99%

Catalog No. AAA1134518
Click to view available options
Quantity:
25 g
50 g
250 g

CAS: 495-40-9 | C10H12O | 148.205 g/mol

Butyrophenone is used as intermediates of Liquid Crystals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyrophenone is used as intermediates of Liquid Crystals

Solubility
Insoluble in water.

Notes
Protect from light. Store under an inert atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-40-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009397
InChI Key FFSAXUULYPJSKH-UHFFFAOYSA-N
Synonym butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
PubChem CID 10315
IUPAC Name 1-phenylbutan-1-one
SMILES CCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 11°C to 13°C
Density 1.021
Boiling Point 222°C
Flash Point 88°C (190°F)
Odor Characteristic
Refractive Index 1.5195
Quantity 50 g
Beilstein 508305
Solubility Information Insoluble in water.
Formula Weight 148.21
Percent Purity 99%
Chemical Name or Material Butyrophenone
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P370+P378q-P374-P501c
EINECSNumber 207-799-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.