Alfa Aesar™ Butyrophenone, 99%

Manufacturer: Alfa Aesar™ A1134514

Quantity 25g

 View more versions of this product

Catalog No. AAA1134514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 495-40-9
    Molecular Formula C10H12O
    Formula Weight 148.21
    MDL Number MFCD00009397
    Beilstein 508305
    Synonym Phenyl n-propyl ketone
    Chemical Name or Material Butyrophenone

    Specifications

    Formula Weight 148.21
    CAS 495-40-9
    Synonym Phenyl n-propyl ketone
    Health Hazard 1 Warning
    Molecular Formula C10H12O
    Beilstein 508305
    Boiling Point 222°C
    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00009397
    Melting Point 11° to 13°C
    Flash Point 88°C (190°F)
    Odor Characteristic
    Chemical Name or Material Butyrophenone
    Assay 99%

    Description

    Applications
    Butyrophenone is used as intermediates of Liquid Crystals

    Solubility
    Insoluble in water.

    Notes
    Protect from light. Store under an inert atmosphere.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit