Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Butyryl-L-carnitine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg
Specifications
Specifications
| CAS | 25576-40-3 |
| Linear Formula | C11H21 NO4 |
| MDL Number | MFCD01674827 |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxobutoxy)-1-propanaminium inner salt; Butanoyl-L-carnitine; C4-Carnitine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 231.29 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction