Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyrylcholine esterase Polyclonal Antibody, Huabio

Catalog No. 502544569
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4569 Supplier HUABIO RESEARCH INC Supplier No. HA500248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Butyrylcholine esterase Polyclonal antibody specifically detects Butyrylcholine esterase in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Butyrylcholine esterase
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene BCHE
Gene Accession No. P06276
Gene Alias acylcholine acylhydrolase; BCHE; BtChE; butylcholinesterase; butyrylcholine esterase; butyrylcholinesterase; C730038G20Rik; CHE1; CHE2; Choline esterase II; cholinesterase; cholinesterase (serum) 2; cholinesterase 1; E1; EC 3.1.1.8; pseudocholinesterase
Host Species Rabbit
Immunogen Recombinant protein within Human Butyrylcholine esterase aa 300-510.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.