Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Butyrylcholinesterase/BCHE Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298275MFV500
Description
Butyrylcholinesterase/BCHE Polyclonal antibody specifically detects Butyrylcholinesterase/BCHE in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Butyrylcholinesterase/BCHE | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
Acylcholine acylhydrolase, Butyrylcholine esterase, butyrylcholinesterase, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase | |
Produced in rabbits immunized with purified, recombinant Mouse Butyrylcholinesterase/BCHE (Accession#: NP_033868.3; Met1-Leu603) | |
0.1 mL | |
Alzheimers Research, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
590 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction