Learn More
Invitrogen™ Butyrylcholinesterase Recombinant Rabbit Monoclonal Antibody (1E9)

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | Butyrylcholinesterase |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 1E9 |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 |
| Gene | BCHE |
| Gene Accession No. | P06276 |
| Gene Alias | acylcholine acylhydrolase; BCHE; BtChE; butylcholinesterase; butyrylcholine esterase; butyrylcholinesterase; C730038G20Rik; CHE1; CHE2; Choline esterase II; cholinesterase; cholinesterase (serum) 2; cholinesterase 1; E1; EC 3.1.1.8; pseudocholinesterase |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.