Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Butyrylcholinesterase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581319 Supplier Invitrogen™ Supplier No. PA581319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse Butyrylcholinesterase/BCHE.

Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Butyrylcholinesterase
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene BCHE
Gene Accession No. Q03311
Gene Alias acylcholine acylhydrolase; BCHE; BtChE; butylcholinesterase; butyrylcholine esterase; butyrylcholinesterase; C730038G20Rik; CHE1; CHE2; Choline esterase II; cholinesterase; cholinesterase (serum) 2; cholinesterase 1; E1; EC 3.1.1.8; pseudocholinesterase
Gene Symbols BCHE
Host Species Rabbit
Immunogen Recombinant Mouse Butyrylcholinesterase/BCHE Protein (Met1-Leu603).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12038
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.