Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Butyrylcholinesterase Recombinant Rabbit Monoclonal Antibody (4N2N9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542897
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa. Immunogen sequence: ANFAKYGNPN ETQNNSTSWP VFKSTEQKYL TLNTESTRIM TKLRAQQCRF WTSFFPKVLE MTGNIDEAEW EWKAGFHRWN NYMMDWKNQF NDYTSKKESC VGL.
Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage.
Specifications
Butyrylcholinesterase | |
Recombinant Monoclonal | |
1.3 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06276 | |
BCHE | |
A synthetic peptide corresponding to a sequence within amino acids 500-602 of human Butyrylcholinesterase (P06276). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
4N2N9 | |
Unconjugated | |
BCHE | |
acylcholine acylhydrolase; BCHE; BtChE; butylcholinesterase; butyrylcholine esterase; butyrylcholinesterase; C730038G20Rik; CHE1; CHE2; Choline esterase II; cholinesterase; cholinesterase (serum) 2; cholinesterase 1; E1; EC 3.1.1.8; pseudocholinesterase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
590 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction