Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUV496 Biosimilar Anti-Human CD52 , BD OptiBuild™
Biosimilar Monoclonal Antibody
Supplier: BD Biosciences 758153
Description
CD52 Monoclonal antibody specifically detects CD52 in Human samples. It is validated for Flow cytometry
Specifications
CD52 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
Alemtuzumab297.rMAb (also known as Alemtuzumab N297A Biosimilar.rMAb) | |
Brilliant Ultraviolet 496 | |
CAMPATH-1 antigen; HE5 | |
Human CD52 | |
50 μg | |
1043 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction