Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUV563 Mouse Anti-Human CD11b (Integrin αM), BD Horizon™

Catalog No. BDB569701 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569701 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569701 Supplier BD Biosciences Supplier No. 569701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11b (Integrin αM) Monoclonal antibody specifically detects CD11b (Integrin αM) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b (Integrin αM)
Applications Flow Cytometry
Classification Monoclonal
Clone ICRF44 (also known as 44)
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 563
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MAC-1A; Mac-1; ITGAM; Integrin alpha M; CR3A; CR-3 alpha; Mo1; SLEB6
Host Species Mouse
Immunogen Human monocytes
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Gene ID (Entrez) 3684
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.