Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUV615 Mouse Anti-Human CD8, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569783
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow cytometry
Specifications
CD8 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
RPA-T8 | |
Brilliant Ultraviolet 615 | |
CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 | |
Human CD8a | |
50 μg | |
925 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction