Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUV615 Mouse Anti-Human Chondroitin Sulfate (NG2), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756718
Description
Chondroitin Sulfate (NG2) Monoclonal antibody specifically detects Chondroitin Sulfate (NG2) in Human samples. It is validated for Flow cytometry
Specifications
| Chondroitin Sulfate (NG2) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
| IgG1 κ |
| Flow Cytometry | |
| 7.1 | |
| Brilliant Ultraviolet 615 | |
| CSPG4; HMW-MAA; MCSP; MCSPG; NG2 | |
| Human BM stromal cell line established by infection with SV-40 | |
| 50 μg | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction