Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUV805 Rat Anti-Mouse CD274 (PD-L1), BD Horizon™

Catalog No. BDB569606 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569606 0.1 mg
BDB569607 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569606 Supplier BD Biosciences Supplier No. 569606
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD274 (PD-L1) Monoclonal antibody specifically detects CD274 (PD-L1) in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD274 (PD-L1)
Applications Flow Cytometry
Classification Monoclonal
Clone MIH5
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 805
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias B7-H1, PD-L1; PD1L1; Programmed death ligand 1
Host Species Rat
Immunogen DBA/2 mouse lymphoma L5178Y transfected with Pdcd1lg1 cDNA
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.