Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUV805 Rat Anti-Mouse IFN-γ, BD Horizon™

Catalog No. BDB569626 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569626 25 μg
BDB569625 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569626 Supplier BD Biosciences Supplier No. 569626
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IFN-γ Monoclonal antibody specifically detects IFN-γ in Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry
Classification Monoclonal
Clone XMG1.2
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 805
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ifg; Ifng; IFN-γ; IFN-g; IFN-gamma; Interferon gamma; Type II Interferon
Host Species Rat
Immunogen Mouse IFN-γ Recombinant Protein
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 15978
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.