Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUV805 Rat Anti-Mouse IL-17A, BD Horizon™
Rat Monoclonal Antibody
$118.00 - $298.00
Specifications
Antigen | IL-17A |
---|---|
Clone | TC11-18H10 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
IL-17A Monoclonal antibody specifically detects IL-17A in Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
IL-17A | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
16171 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
TC11-18H10 | |
Flow Cytometry | |
Brilliant Ultraviolet 805 | |
Rat | |
Mouse | |
IL-17; Il17; IL-17A; Il17a; CTLA-8; Ctla8; Interleukin-17A; interleukin 17A | |
Recombinant Mouse IL-17A Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title