Learn More
BD Veritor™ Buy 2 CLIA waived strep A (256040) and 1 CLIA flu (256045) and receive a BD Veritor analyzer (256066) PROMO
Shop All BD Diagnostic Systems ProductsDescription
256040 - Rapid chromatographic immunoassay for the direct and qualitative detection of Group A Streptococcus antigen from throat swabs of symptomatic patient.
256045 - Chromatographic immunoassay to detect influenza A or B nucleoprotein antigens from respiratory specimens of symptomatic patients within 10 minutes
256066 - Point of Care detection of infectious diseases such as influenza A and B, respiratory syncytial virus (RSV) and group A strep. BD Veritor Plus Analyzer provides lab-quality immunoassay test results within minutes.
Includes:
256066
- Number of reads per instrument: 3500
- Rechargeable lithium battery
- Print-enabled
256040
- BD Veritor System Group A Strep Device (30 devices): Foil pouched device containing one reactive strip. Each strip has one test line of polyclonal antibody specific to Strep A antigen and a positive control line containing purified Strep A antigen
- BD GAS Reagent 1 (Bottle with 4mL reagent): Dilute acetic acid solution
- BD GAS Reagent 2: (30 tubes with 200μL reagent): Sodium nitrite and EDTA
- Individually packaged sterile swabs
- Positive Control Swab
- Negative Control Swab
256045
- Lateral flow test strip
- Flu A+/B- control swab
- Flu B+/A- control swab
- RV reagent C
Specifications
Specifications
| For Use With (Application) | Rapid Diagnostic Testing |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.