Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Biosimilar Anti-Human CD20 Rituximab297, BD Horizon™
Biosimilar Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | CD20 Rituximab297 |
---|---|
Clone | Rituximab297.rMAb (also known as Rituximab N297A Biosimilar.rMAb) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
CD20 Rituximab297 Monoclonal antibody specifically detects CD20 Rituximab297 in Human samples. It is validated for Flow cytometrySpecifications
CD20 Rituximab297 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
B1; Bp35; LEU-16; MS4A1 | |
Human CD20 | |
Affinity Chromatography |
Rituximab297.rMAb (also known as Rituximab N297A Biosimilar.rMAb) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
931 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title