Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human CD77, BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | CD77 |
---|---|
Clone | 5B5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
CD77 Monoclonal antibody specifically detects CD77 in Human samples. It is validated for Flow cytometrySpecifications
CD77 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
BLA; Gb3; Pk blood group antigen; CTH | |
IgM κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
5B5 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Initially Adherent Human Peripheral Blood Cells | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title