Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Mouse Anti-Human Chondroitin Sulfate (NG2), BD Horizon™

Catalog No. BDB570396 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570396 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570396 Supplier BD Biosciences Supplier No. 570396
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chondroitin Sulfate (NG2) Monoclonal antibody specifically detects Chondroitin Sulfate (NG2) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondroitin Sulfate (NG2)
Applications Flow Cytometry
Classification Monoclonal
Clone 7.1
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CSPG4; HMW-MAA; MCSP; MCSPG; NG2
Host Species Mouse
Immunogen Human BM stromal cell line established by infection with SV-40
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Gene ID (Entrez) 1464
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.